To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Ephrin A2
- Antigen Symbol:EFNA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 μg
- Gene ID:1943
- Antigen Synonyms:Epl6|EPLG6|Eplg6|CEK7L|Ephrin-A2|LERK-6|LERK6|Elf1|EPH-related receptor tyrosine kinase ligand 6|HEK7-L|EphrinA2|HEK7 ligand
- Storage Buffer:Phosphate buffered saline, pH 7.2.
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Purification:Immunogen affinity purified
- Cat. No.:89300-180
- Supplier no.:GTX30999
Specifications
About this item
Rabbit polyclonal antibody to Ephrin A2
We recommend the following starting dilutions: Western Blot 1:500-1:1000, ELISA 1:5000-1:10000,. Optimal concentration should be determined experimentally by the end user.
Type: Primary
Antigen: EFNA2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse