To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Solute Carrier Family 6 (Neurotransmitter Transporter), Member 3
- Antigen Symbol:DAT1 (solute carrier Family 6 (neurotransmitter transporter dopamine) member 3)
- Clonality:Monoclonal
- Clone:DAT-Nt(4F8)
- Conjugation:Unconjugated
- Host:Rat
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Monkey,Rat,Mouse
- Western Blot:Yes
- Size:50 μl
- Environmentally Preferable:
- Gene ID:6531
- Antigen Synonyms:DAT1|PKDYS|DAT
- Storage Buffer:Tissue Culture Supernatant
- Immunogen:N-terminus of human dopamine transporter fused to Glutathione S-transferase.
- Purification:Tissue Culture Supernatant
- Cat. No.:89299-080
- Supplier no.:GTX30992
Specifications
About this item
Rat monoclonal antibody [DAT-Nt(4F8)] to DAT1
Recommended Starting Dilutions:Immunohistology - Frozen: 1/1000 - 1/10 000Not tested in other applications. Optimal dilution / concentration should be determined by the end user.
Type: Primary
Antigen: DAT1 (solute carrier Family 6 (neurotransmitter transporter dopamine) member 3)
Clonality: Monoclonal
Clone: DAT-Nt(4F8)
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse, Monkey, Rat