Anti-SATB2 Rabbit Polyclonal Antibody
Catalog # 89300-176
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:SATB Homeobox 2
- Antigen Symbol:SATB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 μg
- Gene ID:23314
- Antigen Synonyms:FLJ21474|MGC119477|MGC119474|SATB family member 2|DNA-binding protein SATB2|FLJ32076|special AT-rich sequence-binding protein 2|KIAA1034
- Storage Buffer:PBS, pH7.2 containing 0.02% sodium azide.
- Storage Temperature:Keep as concentrated solution, aliquot and store at 4°C.
- Concentration:1 mg/ml
- Immunogen:Peptide: 14 amino acids near the carboxy terminus of the human SATB2
- Purification:Affinity purified with antigen
- Cat. No.:89300-176
- Supplier no.:GTX30894
Specifications
About this item
Rabbit polyclonal to SATB2
For ELISA: Use at an assay dependent dilution. For IHC: Use at an assay dependent dilution. For WB: Use at a concetration of 2-4 µg/ml. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: SATB2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat