Anti-HIPK2 Rabbit Polyclonal Antibody
Catalog # 89262-088
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:HomeoDomain Interacting Protein kinase 2
- Antigen Symbol:HIPK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Dog,Rat,Mouse
- Western Blot:Yes
- Size:50 μg
- Gene ID:28996
- Antigen Synonyms:PRO0593|EC 2.7.11.1|HHIPK2
- Storage Buffer:PBS, containing 2% sucrose
- Storage Temperature:Upon Receipt - Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Synthetic peptide – Immunogen sequence information is commercially sensitive
- Purification:Affinity purified with antigen
- Cat. No.:89262-088
- Supplier no.:GTX30803
Specifications
About this item
Rabbit polyclonal to HIPK2
For WB: Use at a concentration of 0.25 ?g/ml. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user
Type: Primary
Antigen: HIPK2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Dog, Rat