Anti-BCAS1 Rabbit Polyclonal Antibody
Catalog # 89262-076
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:NABC1 (breast carcinoma amplified sequence 1)
- Antigen Symbol:BCAS1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:8537
- Antigen Synonyms:NABC1|AIBC1
- Storage Buffer:0.1 mg in 0.1 ml (1 mg/ml) in PBS containing 0.02% sodium azide.
- Storage Temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Reacts with residues 532-550 [DTNSLQNGDKLQKRPEKRQ] of human NABC1.
- Purification:IgG
- Cat. No.:89262-076
- Supplier no.:GTX30622
Specifications
About this item
Rabbit polyclonal antibody to NABC1
For WB: Use at a dilution of 1:500 - 1:1,000. This antibody may be used for Western blot analysis where 2 bands are seen at ~120 and ~80 kDa. NABC1 is a 60 kDa protein; however it forms dimers, which are readily detectable in MCF-7 cells. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.
Type: Primary
Antigen: BCAS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human