Anti-CEACAM5 Chicken Polyclonal Antibody
89261-894
: Genetex
- Antibody Type:Primary
- Antigen Name:Carcinoembryonic Antigen-related Cell Adhesion Molecule 5
- Antigen Symbol:CEACAM5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Chicken
- Isotype:IgY
- Reactivity:Human
- Western Blot:Yes
- Size:50 μg
- Gene ID:1048
- Antigen Synonyms:CEA|CD66e|Carcinoembryonic antigen-related cell adhesion molecule 5|Carcinoembryonic antigen
- Storage Buffer:PBS, no preservatives added
- Storage Temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:Recombinant fragment, corresponding to amino acids 460-600 of Human CEACAM5.
- Purification:Affinity purified with antigen
- Cat. No.:89261-894
- Supplier no.:GTX19128
For WB: Use at an asssay dependent dilution. Predicted molecular weight: 77 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.
Type: Primary
Antigen: CEACAM5
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Chicken
Isotype:
Reactivity: