Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-VAV2 Sheep Polyclonal Antibody
Anti-VAV2 Sheep Polyclonal Antibody
Catalog # 89261-806
Supplier:  Genetex
Anti-VAV2 Sheep Polyclonal Antibody
Catalog # 89261-806
Supplier:  Genetex
Supplier Number:  GTX16136

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Vav 2 Guanine Nucleotide Exchange Factor
  • Antigen Symbol:
    VAV2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Sheep
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    250μg
  • Gene ID:
    7410
  • Antigen Synonyms:
    VAV-2|vav 2 oncogene|guanine nucleotide exchange factor VAV2
  • Storage Buffer:
    Preservative: 0.08% Sodium Azide; Constituents: PBS
  • Storage Temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Immunogen:
    His tagged fusion protein, corresponding to amino acids 287-578 of Human VAV2.
  • Purification:
    IgG fraction
  • Cat. No.:
    89261-806
  • Supplier no.:
    GTX16136

Specifications

About this item

IP: Use at an assay dependent dilution. WB: Use at a dilution of 1/5000. Predicted molecular weight: 101 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: VAV2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Sheep
Isotype:
Reactivity: Human