Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-BAFF Mouse Monoclonal Antibody [clone: Buffy XI]
Anti-BAFF Mouse Monoclonal Antibody [clone: Buffy XI]
Catalog # 89287-962
Supplier:  Genetex
Anti-BAFF Mouse Monoclonal Antibody [clone: Buffy XI]
Catalog # 89287-962
Supplier:  Genetex
Supplier Number:  GTX16087

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    B-cell activating factor
  • Antigen Symbol:
    BAFF
  • Clonality:
    Monoclonal
  • Clone:
    Buffy XI
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    50 μg
  • Gene ID:
    10673
  • Antigen Synonyms:
    CD257|ZTNF4|THANK|TALL-1|TNFSF20|DTL|TNFSF13B|TALL1|TNLG7A|BLYS
  • Storage Buffer:
    Preservative: 0.02% Sodium Azide; Constituents: PBS
  • Storage Temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Concentration:
    1 mg/ml
  • Immunogen:
    Extracellular domain of Human BAFF, corresponding to amino acids 137 - 285.
  • Purification:
    >95% by SDS-PAGE
  • Cat. No.:
    89287-962
  • Supplier no.:
    GTX16087

Specifications

About this item

WB: Use at a concentration of 1 ?g/ml. Predicted molecular weight: 34 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: BAFF
Clonality: Monoclonal
Clone: Buffy XI
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human