Order Entry
United States
ContactUsLinkComponent
Anti-NIFK Mouse Monoclonal Antibody [clone: 18E148]
Anti-NIFK Mouse Monoclonal Antibody [clone: 18E148]
Catalog # 89261-524
Supplier:  Genetex
Anti-NIFK Mouse Monoclonal Antibody [clone: 18E148]
Catalog # 89261-524
Supplier:  Genetex
Supplier Number:  GTX13880
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Nucleolar Protein Interacting With The Fha Domain Of Mki67
  • Antigen Symbol:
    NIFK
  • Clonality:
    Monoclonal
  • Clone:
    18E148
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μg
  • Environmentally Preferable:
  • Gene ID:
    84365
  • Antigen Synonyms:
    MKI67 FHA Domain Interacting Nucleolar Phosphoprotein|MK67I_HUMAN.|MKI67IP|pKi67|NOPP34|hNK|Nucleolar phosphoprotein Nopp34|Nucleolar protein interacting with the FHA domain of pKI 67
  • Storage Buffer:
    Preservative: 0.05% Sodium Azide; Constituents: 0.2% Gelatin, PBS
  • Storage Temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Concentration:
    0.5 mg/ml
  • Immunogen:
    Synthetic peptide, corresponding to amino acids 100-200 of Human NIFK.
  • Purification:
    Protein G purified
  • Cat. No.:
    89261-524
  • Supplier no.:
    GTX13880

Specifications

About this item

WB: Use at a concentration of 0.5 - 3 ?g/ml. Predicted molecular weight: 34 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: NIFK
Clonality: Monoclonal
Clone: 18E148
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human