Anti-PAGA Rabbit Polyclonal Antibody
Catalog # 89280-098
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Anthrax protective Antigen
- Antigen Symbol:PAGA
- Clonality:Polyclonal
- Host:Rabbit
- Western Blot:Yes
- Size:200 µL
- Antigen Synonyms:AnthraxPA
- Cat. No.:89280-098
- Supplier no.:GTX13809
Specifications
About this item
Type: Primary
Antigen: PAGA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: 0