Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-GMNN Rabbit Polyclonal Antibody
Anti-GMNN Rabbit Polyclonal Antibody
  89280-056
 :  Genetex
Anti-GMNN Rabbit Polyclonal Antibody
  89280-056
 :  Genetex
 :  GTX12147

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Gemin 1
  • Antigen Symbol:
    GMNN
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    200μl
  • Gene ID:
    51053
  • Antigen Synonyms:
    SMA 3|SMN-1|telomeric|Survival of motor neuron 1 telomeric|Survival of motor neuron 1|SMA|SMA3|T-BCD541|SMN 1|SMN|SMA 2|Gemin1|SMA1|Gemin-1|SMA2|SMN2|SMN_HUMAN.|Component of gems 1|Gemin 1|Survival motor neuron protein|SMN_HUMAN|SMA4|SMA 1|SMA 4|SMN1|survival of motor neuron 1|SMNT|BCD541
  • Storage Buffer:
    Preservative: 0.02% Sodium Azide; Constituents: 1% BSA, PBS. pH 7.4
  • Storage Temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Concentration:
    0.1 mg/ml
  • Immunogen:
    Synthetic peptide: SSENKNLGGVTQES-C conjugated to KLH, corresponding to amino acids 72-85 of Human Geminin.
  • Purification:
    Immunogen affinity purified
  • Cat. No.:
    89280-056
  • Supplier no.:
    GTX12147

 

 

WB: Use at a dilution of 1/500. Detects a band of approximately 33 kDa (predicted molecular weight: 23.5 kDa). Can be blocked with GTX12408 Geminin peptide . Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: GMNN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human