Anti-Ifng Rat Monoclonal Antibody [clone: F1]
Catalog # 89291-874
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Interferon gamma (interferon gamma)
- Antigen Symbol:Ifng
- Clonality:Monoclonal
- Clone:F1
- Conjugation:Unconjugated
- Host:Rat
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Mouse
- Size:50 μg
- Gene ID:15978
- Storage Buffer:Phosphate buffered saline, containing 0.02% sodium azide and 0.1% BSA
- Storage Temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:0.1 mg/ml
- Immunogen:Recombinant full length protein (Mouse).
- Purification:Protein G affinity purified
- Cat. No.:89291-874
- Supplier no.:GTX11855
Specifications
About this item
For IA: Use at an assay dependent dilution. For IP: Use at an assay dependent dilution. For P: Use at an assay dependent dilution. Is unsuitable for Neutralising. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: Ifng
Clonality: Monoclonal
Clone: F1
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse