Anti-APOBEC3F Rabbit Polyclonal Antibody
Catalog # 89323-308
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Apolipoprotein B mRNA Editing Enzyme, Catalytic Polypeptide-Like 3F
- Antigen Symbol:APOBEC3F
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:200316
- Antigen Synonyms:KA6|ARP8|A3F|BK150C2.4.MRNA
- Storage Buffer:0.1M Tris, 0.1M Glycine, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Storage Temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:0.69 mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human APOBEC3F. The exact sequence is proprietary.
- Purification:Purified by antigen-affinity chromatography.
- Cat. No.:89323-308
- Supplier no.:GTX118217
Specifications
About this item
Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: APOBEC3F
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human