Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-RPRD1A Rabbit Polyclonal Antibody
  89322-600
 :  Genetex
Anti-RPRD1A Rabbit Polyclonal Antibody
  89322-600
 :  Genetex
 :  GTX116296

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Regulation Of Nuclear Pre-MRNA Domain Containing 1A 
  • Antigen Symbol:
    RPRD1A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Gene ID:
    55197
  • Antigen Synonyms:
    P15RS|P15INK4B-Related Protein
  • Storage Buffer:
    0.1M Tris, 0.1M Glycine, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
  • Storage Temperature:
    Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
  • Concentration:
    0.69 mg/ml
  • Immunogen:
    Recombinant protein encompassing a sequence within the center region of human P15RS. The exact sequence is proprietary.
  • Purification:
    Purified by antigen-affinity chromatography.
  • Cat. No.:
    89322-600
  • Supplier no.:
    GTX116296

 

 

Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: RPRD1A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human