Anti-PPP2R5A Rabbit Polyclonal Antibody
Catalog # 89312-868
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Protein Phosphatase 2, Regulatory Subunit 5A
- Antigen Symbol:PPP2R5A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Pig,Human,Bovine,Dog,Rat
- Western Blot:Yes
- Size:100 μl
- Gene ID:25970
- Antigen Synonyms:B' alpha isoform|B subunit|B56 alpha isoform|56 kDa regulatory subunit|PR61 alpha isoform|MGC131915|MGC131915 |regulatory subunit B'|R5 alpha isoform|regulatory subunit B (B56)|B56A|PP2A|alpha isoform|PR61A|serine/threonine protein phosphatase 2A|protein phosphatase 2
- Storage Buffer:0.1M Tris, 0.1M Glycine, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Storage Temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human SH2B. The exact sequence is proprietary.
- Purification:Purified by antigen-affinity chromatography.
- Cat. No.:89312-868
- Supplier no.:GTX115780
Specifications
About this item
Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: PPP2R5A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Bovine, Dog, Pig, Rat