Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-AOC3 Rabbit Polyclonal Antibody
Anti-AOC3 Rabbit Polyclonal Antibody
Catalog # 89321-814
Supplier:  Genetex
Anti-AOC3 Rabbit Polyclonal Antibody
Catalog # 89321-814
Supplier:  Genetex
Supplier Number:  GTX114707

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Amine Oxidase, Copper Containing 3
  • Antigen Symbol:
    AOC3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Gene ID:
    5928
  • Antigen Synonyms:
    VAP-1.|Vascular adhesion protein 1|Semicarbazide-sensitive amine oxidase|Membrane primary amine oxidase|Membrane copper amine oxidase|Semicarbazide sensitive amine oxidase|Vascular Adhesion Protein 1|HPAO|SSAO|AOC3|VP97|AOC3_HUMAN|Amine Oxidase|Copper Containing 3 (vascular adhesion protein 1)|Copper amine oxidase
  • Storage Buffer:
    0.1M Tris, 0.1M Glycine, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
  • Storage Temperature:
    Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
  • Concentration:
    0.34 mg/ml
  • Immunogen:
    Recombinant protein encompassing a sequence within the center region of human RbAp48. The exact sequence is proprietary.
  • Purification:
    Purified by antigen-affinity chromatography.
  • Cat. No.:
    89321-814
  • Supplier no.:
    GTX114707

Specifications

About this item

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: AOC3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat