Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-GABPB2 Rabbit Polyclonal Antibody
Anti-GABPB2 Rabbit Polyclonal Antibody
Catalog # 89321-720
Supplier:  Genetex
Anti-GABPB2 Rabbit Polyclonal Antibody
Catalog # 89321-720
Supplier:  Genetex
Supplier Number:  GTX114533

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    GA Binding Protein Transcription Factor, Beta Subunit 2
  • Antigen Symbol:
    GABPB2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Dog
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Gene ID:
    4046
  • Antigen Synonyms:
    BABPB2|beta subuni|E4TF1-53|E4TF1|E4TF1B|NRF2B2|E4TF1-47|beta subunit 1|GA binding protein transcription factor|53kDa|GABPB1|GABPB|beta subunit 2|GABPB2|NRF2B1
  • Storage Buffer:
    0.1M Tris, 0.1M Glycine, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
  • Storage Temperature:
    Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
  • Concentration:
    0.97 mg/ml
  • Immunogen:
    Recombinant protein encompassing a sequence within the center region of human LSP1. The exact sequence is proprietary.
  • Purification:
    Purified by antigen-affinity chromatography.
  • Cat. No.:
    89321-720
  • Supplier no.:
    GTX114533

Specifications

About this item

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: LSP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human