To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Calcium Channel, Voltage-dependent, alpha 2/delta subunit 1
- Antigen Symbol:CACNA2D1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Dog,Rat,Mouse
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:3866
- Antigen Synonyms:CACNA2D1|Voltage gated calcium channel subunit alpha 2/delta 1|Voltage-dependent calcium channel subunit delta-1|MHS3|L type calcium channel subunit alpha 2|CACNL2A|MHS 3|CA2D1_HUMAN|Voltage dependent calcium channel subunit alpha 2/delta 1|CCHL2A|CACN A2|H_DJ0560O14.1|CACNA2|Calcium channel L type DHPR alpha 2 subunit|Calcium channel voltage dependent alpha 2/delta subunit 1|Dihydropyridine sensitive L type calcium channel alpha 2/delta subunit|Dihydropyridine sensitive L type calcium channel subunits alpha 2/delta|WUGSC:H_DJ0560O14.1.|Dihydropyridine receptor alpha 2 subunit|Calcium channel L type alpha 2 polypeptide|Voltage-gated calcium channel subunit alpha-2/delta-1
- Storage Buffer:0.1M Tris, 0.1M Glycine, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Storage Temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:0.98 mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human Cytokeratin 15. The exact sequence is proprietary.
- Purification:Purified by antigen-affinity chromatography.
- Cat. No.:89321-708
- Supplier no.:GTX114517
Specifications
About this item
Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: CACNA2D1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Dog, Rat