Anti-POU2F3 Rabbit Polyclonal Antibody
Catalog # 89320-912
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:POU Class 2 Homeobox 3
- Antigen Symbol:POU2F3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Bovine,Dog,Rat
- Western Blot:Yes
- Size:100 μl
- Gene ID:26135
- Antigen Synonyms:POU transcription factor|MGC126698|Skn-1a|class 2|Epoc-1|OCT11|POU class 2 homeobox 3|PLA-1|transcription factor 3|POU domain|POU2F3|FLJ40063
- Storage Buffer:0.1M Tris, 0.1M Glycine, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Storage Temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human SERBP1. The exact sequence is proprietary.
- Purification:Purified by antigen-affinity chromatography.
- Cat. No.:89320-912
- Supplier no.:GTX112129
Specifications
About this item
Rabbit Polyclonal antibody to SERBP1 (SERPINE1 mRNA binding protein 1)
Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: POU2F3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Bovine, Dog, Rat