Anti-UCHL1 Rabbit Polyclonal Antibody
89320-892
: Genetex
- Antibody Type:Primary
- Antigen Name:Ubiquitin Carboxyl-Terminal Esterase L1
- Antigen Symbol:UCHL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Pig,Rabbit,Human,Bovine,Dog,Rat
- Western Blot:Yes
- Size:100 μl
- Gene ID:56940
- Antigen Synonyms:Neuron cytoplasmic protein 9.5|Ubiquitin thioesterase L1.|PGP9.5|Ubiquitin carboxyl-terminal|hydrolase isozyme L1|Ubiquitin thioesterase L1|UCH-L1|PGP 9.5|PARK5
- Storage Buffer:1XPBS, 1% BSA, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Storage Temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:0.83mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human DUSP22. The exact sequence is proprietary.
- Purification:Purified by antigen-affinity chromatography.
- Cat. No.:89320-892
- Supplier no.:GTX112048
Rabbit Polyclonal antibody to DUSP22 (dual specificity phosphatase 22)
Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: UCHL1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Bovine, Dog, Pig, Rabbit, Rat