Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-PRKAA2 Rabbit Polyclonal Antibody
Anti-PRKAA2 Rabbit Polyclonal Antibody
  89320-874
 :  Genetex
Anti-PRKAA2 Rabbit Polyclonal Antibody
  89320-874
 :  Genetex
 :  GTX112011

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Protein Kinase AMP-activated Catalytic Subunit Alpha 2
  • Antigen Symbol:
    PRKAA2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Pig,
    Human,
    Bovine,
    Dog,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Gene ID:
    221823
  • Antigen Synonyms:
    AMPKa2|AMPK|AMPK2|PRKAA
  • Storage Buffer:
    0.1M Tris, 0.1M Glycine, 10% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
  • Storage Temperature:
    Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
  • Concentration:
    1mg/ml
  • Immunogen:
    Full length human PRPS1L1 Recombinant protein.
  • Purification:
    Purified by antigen-affinity chromatography.
  • Cat. No.:
    89320-874
  • Supplier no.:
    GTX112011

 

 

Rabbit Polyclonal antibody to PRPS1L1 (phosphoribosyl pyrophosphate synthetase 1-like 1)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PRKAA2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Bovine, Dog, Pig, Rat