Anti-ANKRD26 Rabbit Polyclonal Antibody
Catalog # 89320-298
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Ankyrin Repeat Domain 26
- Antigen Symbol:ANKRD26
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:1845
- Antigen Synonyms:VHR
- Storage Buffer:0.1M Tris, 0.1M Glycine, 10% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Storage Temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:0.58mg/ml
- Immunogen:Full length human DUSP3 Recombinant protein.
- Purification:Purified by antigen-affinity chromatography.
- Cat. No.:89320-298
- Supplier no.:GTX109149
Specifications
About this item
Rabbit Polyclonal antibody to DUSP3 (dual specificity phosphatase 3)
Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: ANKRD26
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human