Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-ZC3H12B Rabbit Polyclonal Antibody
Anti-ZC3H12B Rabbit Polyclonal Antibody
Catalog # 89319-760
Supplier:  Genetex
undefined
Anti-ZC3H12B Rabbit Polyclonal Antibody
Catalog # 89319-760
Supplier:  Genetex
Supplier Number:  GTX106952

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Zinc Finger CCCH-type Containing 12B
  • Antigen Symbol:
    ZC3H12B
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Gene ID:
    27339
  • Antigen Synonyms:
    CXorf32|MCP-induced protein 2|MCPIP2|ZC3H12B|Probable ribonuclease ZC3H12B
  • Storage Buffer:
    0.1M Tris, 0.1M Glycine, 10% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
  • Storage Temperature:
    Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
  • Concentration:
    1.04mg/ml
  • Immunogen:
    Recombinant protein encompassing a sequence within the center region of human PRP19. The exact sequence is proprietary.
  • Purification:
    Purified by antigen-affinity chromatography.
  • Cat. No.:
    89319-760
  • Supplier no.:
    GTX106952

Specifications

About this item

Rabbit Polyclonal antibody to PRP19 (PRP19/PSO4 pre-mRNA processing factor 19 homolog (S. cerevisiae))

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: ZC3H12B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat