Anti-LIN7B Rabbit Polyclonal Antibody
Catalog # 89319-116
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Lin-7 homolog B
- Antigen Symbol:LIN7B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:7318
- Antigen Synonyms:MALS-2|LIN-7B|VELI2|MALS2
- Storage Buffer:0.1M Tris, 0.1M Glycine, 10% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Storage Temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the N-terminus region of human UBE1L. The exact sequence is proprietary.
- Purification:Purified by antigen-affinity chromatography.
- Cat. No.:89319-116
- Supplier no.:GTX104103
Specifications
About this item
Rabbit Polyclonal antibody to UBE1L (ubiquitin-like modifier activating enzyme 7)
Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: LIN7B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human