Anti-PIK3AP1 Rabbit Polyclonal Antibody
89318-856
: Genetex
- Antibody Type:Primary
- Antigen Name:Phosphoinositide-3-Kinase Adaptor Protein 1
- Antigen Symbol:PIK3AP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:2551
- Antigen Synonyms:BCAP
- Storage Buffer:0.1M Tris, 0.1M Glycine, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Storage Temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1.02mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the N-terminus region of human GABPA. The exact sequence is proprietary.
- Purification:Purified by antigen-affinity chromatography.
- Cat. No.:89318-856
- Supplier no.:GTX102830
Rabbit Polyclonal antibody to GABPA (GA binding protein transcription factor, alpha subunit 60kDa)
Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: PIK3AP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human