Anti-PRKD2 Rabbit Polyclonal Antibody
89318-782
: Genetex
- Antibody Type:Primary
- Antigen Name:Protein Kinase D2
- Antigen Symbol:PRKD2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:2551
- Antigen Synonyms:PRKCM|HSPC187|Protein kinase D|PKCu|Serine/threonine-protein kinase D2|nPKC-D2|nPKC-D1|Protein kinase C nu type|PKC nu|PRKCN|PKC-u|PKC-nu|PKC mu|PKD|nPKC-nu|PKC-v|PKD3|Serine/threonine-protein kinase D3|PRKD2|Serine/threonine-protein kinase D1|PKCv|PKD1|Protein kinase C mu type|EPK2|PKCmu|PKC v|PKC u|PRKD3|Protein kinase EPK2|PRKD1|PKD2|PKCnu|nPKC-mu
- Storage Buffer:1XPBS, 20% Glycerol (pH7). 0.025% ProClin 300 was added as a preservative.
- Storage Temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:0.62mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human GABPA. The exact sequence is proprietary.
- Purification:Purified by antigen-affinity chromatography.
- Cat. No.:89318-782
- Supplier no.:GTX102572
Rabbit Polyclonal antibody to GABPA (GA binding protein transcription factor, alpha subunit 60kDa)
Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: PRKD2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human