Order Entry
United States
ContactUsLinkComponent
 
Anti-SNX9 Rabbit Polyclonal Antibody
  89318-612
 :  Genetex
undefined
Anti-SNX9 Rabbit Polyclonal Antibody
  89318-612
 :  Genetex
 :  GTX101975
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Sorting Nexin 9
  • Antigen Symbol:
    SNX9
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Dog,
    Rat
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Environmentally Preferable:
  • Gene ID:
    171017
  • Antigen Synonyms:
    MSTP155|Wiskott-Aldrich syndrome protein (WASP) interactor protein|MST155|WISP|SNX9|sorting nexin 9 |SH3 and PX domain-containing protein SH3PX1|SH3PX1|SDP1|RP11-266C7.10|OTTHUMP00000040083
  • Storage Buffer:
    1XPBS, 1% BSA, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
  • Storage Temperature:
    Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
  • Concentration:
    1mg/ml
  • Immunogen:
    Recombinant protein encompassing a sequence within the N-terminus region of human ZNF384. The exact sequence is proprietary.
  • Purification:
    Purified by antigen-affinity chromatography.
  • Cat. No.:
    89318-612
  • Supplier no.:
    GTX101975

 

 

Rabbit Polyclonal antibody to ZNF384 (zinc finger protein 384)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: SNX9
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Dog, Rat