Anti-EhpB6 Mouse Monoclonal Antibody [clone: 1E11]
Catalog # 89317-170
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Pig Transmissible Gastroenteritis Coronavirus
- Antigen Symbol:EhpB6
- Clonality:Monoclonal
- Clone:1E11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:500 μg
- Storage Buffer:PBS (pH 7.4) with 0.09% Sodium Azide
- Storage Temperature:Keep as concentrated solution, aliquot and store at 4°C. We recommend to parafilm vials during storage to prevent evaporation of buffer.
- Concentration:2.6 mg/ml
- Immunogen:Pig Transmissible Gastroenteritis Coronavirus
- Purification:Protein G purified
- Cat. No.:89317-170
- Supplier no.:GTX10024
Specifications
About this item
Mouse monoclonal [1E11] to Pig Transmissible Gastroenteritis Virus
ELISA: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: EhpB6
Clonality: Monoclonal
Clone: 1E11
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human