Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-AFF4 Rabbit Polyclonal Antibody
Catalog # 89398-908
Supplier:  Abgent
undefined
Anti-AFF4 Rabbit Polyclonal Antibody
Catalog # 89398-908
Supplier:  Abgent
Supplier Number:  AP9688A

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    AF4/FMR2 Family, Member 4
  • Antigen Symbol:
    AFF4
  • Clonality:
    Polyclonal
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Antigen Synonyms:
    MCEF|AF5Q31|CHOPS
  • Cat. No.:
    89398-908
  • Supplier no.:
    AP9688A

Specifications

About this item

Type: Primary
Antigen: AFF4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human