Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Anti-IL9 Rabbit Polyclonal Antibody

Anti-IL9 Rabbit Polyclonal Antibody

100 µg
Catalog # 89397-388
Supplier:   Abgent
Supplier #:  AP8934C
Anti-IL9 Rabbit Polyclonal Antibody
Catalog # 89397-388
Supplier:  Abgent
Anti-IL9 Rabbit Polyclonal Antibody

100 µg

Price
$610.38
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Type: Primary
Antigen: IL9
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Documents

Specifications
  • Size:
    100 µg
  • Cat. No.:
    89397-388
  • Clonality:
    Polyclonal
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Antibody Type:
    Primary
  • Antigen Symbol:
    IL9
  • Host:
    Rabbit
  • Antigen Synonyms:
    Interleukin-9|IL-9|IL-9 |Cytokine P40|HP40|P40
  • Antigen Name:
    Interleukin 9
  • ELISA:
    Yes