Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Anti-PLCL1 Rabbit Polyclonal Antibody

Anti-PLCL1 Rabbit Polyclonal Antibody

100 µg
Catalog # 89397-218
Supplier:   Abgent
Supplier #:  AP8850A
Anti-PLCL1 Rabbit Polyclonal Antibody
Catalog # 89397-218
Supplier:  Abgent
Anti-PLCL1 Rabbit Polyclonal Antibody
Anti-PLCL1 Rabbit Polyclonal Antibody

100 µg

Price
$640.88
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Type: Primary
Antigen: PLCL1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Documents

Specifications
  • Size:
    100 µg
  • Cat. No.:
    89397-218
  • Clonality:
    Polyclonal
  • Flow Cytometry:
    Yes
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Antibody Type:
    Primary
  • Antigen Symbol:
    PLCL1
  • Host:
    Rabbit
  • Antigen Synonyms:
    PRIP|PLCL|PLDL1|PPP1R127|PLCE
  • Antigen Name:
    Phospholipase C-like 1
  • ELISA:
    Yes
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.