Anti-DAZL Rabbit Polyclonal Antibody
Catalog # 89395-032
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:Deleted in Azoospermia-like
- Antigen Symbol:DAZL
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Antigen Synonyms:MGC26406|deleted in azoospermia-like |DAZLA|DAZL|HGNC:2685|spermatogenesis gene on the Y-like autosomal|deleted in azoospermia-like autosomal|DAZH|SPGYLA|germline specific RNA binding protein|DAZL1
- Cat. No.:89395-032
- Supplier no.:AP7739D
Specifications
About this item
Type: Primary
Antigen: DAZL
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse