Anti-NAE1 Rabbit Polyclonal Antibody
Catalog # 89393-558
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:NEDD8 Activating Enzyme E1 Subunit 1
- Antigen Symbol:NAE1
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Antigen Synonyms:APPBP1|A-116A10.1|ula-1|HPP1
- Cat. No.:89393-558
- Supplier no.:AP7273E
Specifications
About this item
Type: Primary
Antigen: NAE1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human