Anti-HMGCS1 Rabbit Polyclonal Antibody
Catalog # 89391-788
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:3-hydroxy-3-methylglutaryl-CoA Synthase 1 (soluble)
- Antigen Symbol:HMGCS1
- Clonality:Polyclonal
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Cat. No.:89391-788
- Supplier no.:AP6652C
Specifications
About this item
Type: Primary
Antigen: HMGCS1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse