Anti-GPRASP1 Rabbit Polyclonal Antibody
Catalog # 89390-372
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:G Protein-coupled Receptor Associated Sorting Protein 1
- Antigen Symbol:GPRASP1
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Antigen Synonyms:GASP-1|GASP1|G-protein coupled receptor-associated sorting protein 1|GASP
- Cat. No.:89390-372
- Supplier no.:AP5831B
Specifications
About this item
Type: Primary
Antigen: GPRASP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human