Anti-PYROXD1 Rabbit Polyclonal Antibody
Catalog # 89390-306
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:Pyridine Nucleotide-disulphide Oxidoreductase Domain 1
- Antigen Symbol:PYROXD1
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Antigen Synonyms:pyridine nucleotide-disulfide oxidoreductase domain-containing protein 1|pyridine nucleotide-disulphide oxidoreductase domain 1|FLJ22028
- Cat. No.:89390-306
- Supplier no.:AP5795C
Specifications
About this item
Type: Primary
Antigen: PYROXD1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human