Anti-HMGCLL1 Rabbit Polyclonal Antibody
89390-250
: Abgent
- Antibody Type:Primary
- Antigen Name:3-hydroxymethyl-3-methylglutaryl-CoA lyase-like 1
- Antigen Symbol:HMGCLL1
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Cat. No.:89390-250
- Supplier no.:AP5765C
Type: Primary
Antigen: HMGCLL1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse