Anti-RSAD1 Rabbit Polyclonal Antibody
Catalog # 89389-256
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:Radical S-adenosyl Methionine Domain Containing 1
- Antigen Symbol:RSAD1
- Clonality:Polyclonal
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cat. No.:89389-256
- Supplier no.:AP5262B
Specifications
About this item
Type: Primary
Antigen: RSAD1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human