Anti-HVCN1 Rabbit Polyclonal Antibody
Catalog # 89388-812
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:Hydrogen Voltage-Gated Channel 1
- Antigen Symbol:HVCN1
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Antigen Synonyms:Hydrogen voltage-gated channel 1|HVCN1|HV1|Voltage-gated hydrogen channel 1|VSOP|UNQ578/PRO1140
- Cat. No.:89388-812
- Supplier no.:AP5042A
Specifications
About this item
Type: Primary
Antigen: HVCN1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human