Anti-MECP2 Rabbit Polyclonal Antibody
Catalog # 89386-056
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:Methyl CPG Binding Protein 2
- Antigen Symbol:MECP2
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Antigen Synonyms:RTS|MRX79|AUTSX3|MRX16|PPMX|RS|MRXS13|MRXSL|RTT
- Cat. No.:89386-056
- Supplier no.:AP2545D
Specifications
About this item
Type: Primary
Antigen: MECP2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse