Anti-STAM2 Rabbit Polyclonal Antibody
100 µg
Catalog # 89383-958
Supplier: Abgent
Supplier #: AP14042C
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Type: Primary
Antigen: STAM2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Documents
Specifications
- Size:100 µg
- Cat. No.:89383-958
- Clonality:Polyclonal
- Reactivity:Human
- Western Blot:Yes
- Antibody Type:Primary
- Antigen Symbol:STAM2
- Host:Rabbit
- Antigen Synonyms:HGNC:11358|STAM-like protein containing SH3 and ITAM domains 2|DKFZp564C047|signal transducing adaptor molecule 2|STAM2B |signal transducing adaptor molecule (SH3 domain and ITAM motif) 2 |STAM2A|Hbp|STAM2
- Antigen Name:Signal Transducing Adaptor Molecule (SH3 Domain And ITAM motif) 2
- ELISA:Yes
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



