Anti-DCBLD2 Rabbit Polyclonal Antibody
89383-518
: Abgent
- Antibody Type:Primary
- Antigen Name:Discoidin, CUB And LCCL Domain Containing 2
- Antigen Symbol:DCBLD2
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Antigen Synonyms:ESDN|coagulation factor V/VIII-homology domains protein 1|endothelial and smooth muscle cell-derived neuropilin-like protein|CLCP1|DCBD2_HUMAN.|LCCL and coagulation factor V/VIII-homology domains protein 1|Discoidin|CUB and LCCL domain-containing protein 2|CUB
- Cat. No.:89383-518
- Supplier no.:AP13827B
Type: Primary
Antigen: DCBLD2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse