Anti-PARS2 Rabbit Polyclonal Antibody
Catalog # 89378-464
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:Prolyl-tRNA Synthetase 2, mitochondrial
- Antigen Symbol:PARS2
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Cat. No.:89378-464
- Supplier no.:AP11090B
Specifications
About this item
Type: Primary
Antigen: PARS2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse