Anti-TCL6 Rabbit Polyclonal Antibody
Catalog # 89377-948
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Symbol:TCL6
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cat. No.:89377-948
- Supplier no.:AP10849B
Specifications
About this item
Type: Primary
Antigen: TCL6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human