Anti-LARP6 Rabbit Polyclonal Antibody
Catalog # 89377-904
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:La ribonucleoprotein Domain Family member 6
- Antigen Symbol:LARP6
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Antigen Synonyms:ACHN
- Cat. No.:89377-904
- Supplier no.:AP10830A
Specifications
About this item
Type: Primary
Antigen: LARP6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human