Anti-VGLL2 Rabbit Polyclonal Antibody
Catalog # 89377-892
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:Vestigial-like Family Member 2
- Antigen Symbol:VGLL2
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Antigen Synonyms:VITO1|VGL2
- Cat. No.:89377-892
- Supplier no.:AP10824A
Specifications
About this item
Type: Primary
Antigen: VGLL2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse