Anti-KIAA1310 Rabbit Polyclonal Antibody
89377-726
: Abgent
- Antibody Type:Primary
- Antigen Symbol:KIAA1310
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cat. No.:89377-726
- Supplier no.:AP10747C
Type: Primary
Antigen: KIAA1310
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human