Anti-NKX2-1 Rabbit Polyclonal Antibody
Catalog # 89377-696
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:NK2 homeobox 1
- Antigen Symbol:NKX2-1
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Antigen Synonyms:TTF-1|BCH|NKX2A|TEBP|NKX2.1|TTF1|BHC|NK-2|T/EBP|TITF1
- Cat. No.:89377-696
- Supplier no.:AP10734A
Specifications
About this item
Type: Primary
Antigen: NKX2-1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human