Anti-CIC Rabbit Polyclonal Antibody
89377-634
: Abgent
- Antibody Type:Primary
- Antigen Symbol:CIC
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Cat. No.:89377-634
- Supplier no.:AP10705A
Type: Primary
Antigen: CIC
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: