Anti-CHST2 Rabbit Polyclonal Antibody
Catalog # 89377-632
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:Carbohydrate Sulfotransferase 2
- Antigen Symbol:CHST2
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Antigen Synonyms:C6ST|glcNAc6ST-1|Gn6ST-1|HEL-S-75|GST2|GST-2
- Cat. No.:89377-632
- Supplier no.:AP10704C
Specifications
About this item
Type: Primary
Antigen: CHST2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: